Anti-CD3 Rat Monoclonal Antibody (BG Violet 450) [clone: 17A2]

Supplier: Biogems
05112-40-25
75842-894EA 96.79 USD
75842-894
Anti-CD3 Rat Monoclonal Antibody (BG Violet 450) [clone: 17A2]
Antibodies
The 17A2 monoclonal antibody specifically reacts with the mouse T lymphocytes receptor (TCR) associated CD3 complex, resulting in cellular activation and proliferation. CD3 is expressed by thymocytes and mature lymphocytes, and contains γ, δ, and ε subunits, involved in the assembly, trafficking, and surface expression of T-cell receptor complex. The interaction between the T lymphocytes and the 17A2 antibody can be blocked by the 145-2C11 anti-CD3e antibody, demonstrating that the 17A2 antibody recognizes the CD3 ε chain.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR