Anti-ITGB3 mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: VI-PL2]

Supplier: BIOGEMS INTERNATIONAL INC.
03711-50-100
75842-884EA 410.48 USD
75842-884
Anti-ITGB3 mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: VI-PL2]
Antibodies
The VI-PL2 monoclonal antibody reacts with CD61, a type I integral transmembrane glycoprotein also known as integrin beta 3. CD61 forms a complex with CD41 (platelet gpIIb) and CD51 (integrin alphaV). It is expressed on osteoclasts, fibroblasts, platelets, macrophages, and megakaryocytes and appears to bind to fibronectin, vitronectin, thrombospondin, and fibrinogen and vWF. The VI-PL2 antibody is reported to be cross-reactive with baboon, rhesus, and cynomolgus.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: VI-PL2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR