Anti-CD41 Mab 100ug
Supplier: Biogems
Anti-CD41, Monoclonal antibody, Clone: MWReg30, Host: Rat, Species: Mouse, Isotype: IgG1, kappa, Conjugate: FITC, Formulation: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. FC, Size: 100 ug