Anti-PECAM1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 390]

Supplier: BIOGEMS INTERNATIONAL INC.
03412-80-100
75842-854EA 677.69 USD
75842-854
Anti-PECAM1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 390]
Antibodies
The 390 monoclonal antibody specifically reacts with mouse cd31, a 130-140 kDA type I transmembrane glycoprotein also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1). CD31 is reported to bind to CD38 and is expressed on platelets, monocytes, granulocytes, and endothelial cells. It plays a role in angiogenesis, wound healing, cellular migration, and signal transduction.

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 390
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR