Anti-PECAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 390]

Supplier: BIOGEMS INTERNATIONAL INC.
03412-77-100
75842-850EA 359.69 USD
75842-850
Anti-PECAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 390]
Antibodies
The 390 monoclonal antibody specifically reacts with mouse cd31, a 130-140 kDA type I transmembrane glycoprotein also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1). CD31 is reported to bind to CD38 and is expressed on platelets, monocytes, granulocytes, and endothelial cells. It plays a role in angiogenesis, wound healing, cellular migration, and signal transduction.

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 390
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR