Anti-CD31 Mab 200ug
Supplier: Biogems
Anti-CD31 Monoclonal antibody, Clone: 390, Host: Rat, Species: Mouse, Isotype: IgG2a, kappa, Conjugate: PE, Formulation: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. Application: FC, Size: 200ug