Anti-CD21 mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB5]

Supplier: BIOGEMS INTERNATIONAL INC.
03311-60-100
75842-842EA 530.77 USD
75842-842
Anti-CD21 mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB5]
Antibodies
The HB5 monoclonal antibody specifically reacts with human CD21, a 145 kDA glycosylated type I integral membrane protein. CD21 is reported to be involved in B cell activation and proliferation. It is expressed on follicular dendritic cells, mature B cells, a subset of T cells, and a subset of epithelial cells. It is the receptor utilized by the Epstein-Barr virus to infect B cells.

Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: HB5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR