Anti-CD11c Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3.9]

Supplier: BIOGEMS INTERNATIONAL INC.
03231-50-25
75842-820EA 96.79 USD
75842-820
Anti-CD11c Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3.9]
Antibodies
The 3.9 monoclonal antibody specifically binds to the human adhesion glycoprotein CD11c, a 150 kDA integrin α chain also known as integrin alpha X. It is expressed on macrophages, granulocytes, monocytes, dendritic cells, natural killer cells, and subsets of B and T lymphocytes. The CD11c/CD18 complex associates with the iC3b, fibrinogen and ICAM-1 and has an important function in leukocyte adhesion.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR