Anti-CD11a Rat Monoclonal Antibody [clone: M17/4]

Supplier: BIOGEMS INTERNATIONAL INC.
03222-25-50
75842-808EA 198.24 USD
75842-808
Anti-CD11a Rat Monoclonal Antibody [clone: M17/4]
Antibodies
The M17/4 monoclonal antibody specifically reacts with mouse CD11a, a 180kDA glycoprotein. CD11a is the integrin alpha L chain that forms with CD18 the heterodimer molecule Lymphocyte Function-associated Antigen-1 (LFA-1). LFA-1 is expressed on all leukocytes and is involved in intercellular adhesions. The M17/4 antibody blocks many of the LFA-1 mediated cell interaction and in vivo it prolongs allograft survival, inhibits the autoimmune response, and reduces graft versus host reactions.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: M17/4
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR