Anti-CD11b (M1/70) Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: M1/70]

Supplier: BIOGEMS INTERNATIONAL INC.
03221-70-25
75842-792EA 90.32 USD
75842-792
Anti-CD11b (M1/70) Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: M1/70]
Antibodies
The M1/70 monoclonal antibody specifically reacts with the 170 kDa αM integrin chain of mouse CD11b from the Mac-1 integrin (CD11b/CD18). Mac-1 binds to C3bi, CD54 (ICAM-1), and fibrinogen, and it is expressed by granulocytes, macrophages, NK cells, myeloid-derived dendritic cells, microglia, activated lymphocytes, and mouse B-1 cells. The expression is up-regulated on activated neutrophils at the same time that L-selectin is shed from the cell surface. The M1/70 antibody is used for the detection of monocytes, granulocytes, and a subset of natural killer cells in human peripheral blood.M1/70 blocks C3bi binding and cell adherence, but not cell-mediated lysis and it cross-reacts with human CD11b.

Type: Primary
Antigen: CD11b (M1/70)
Clonality: Monoclonal
Clone: M1/70
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR