Anti-CD90.2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 30-H12]

Supplier: BIOGEMS INTERNATIONAL INC.
03012-50-100
75842-784EA 96.79 USD
75842-784
Anti-CD90.2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 30-H12]
Antibodies
The 30-H12 monoclonal antibody specifically binds to mouse CD90.2, an alloantigen known as Thy-1.2, expressed on thymocytes, mature T cells, epithelial cells, neurons, hematopoietic stem cells, and fibroblasts. CD90 is a membrane glycoprotein that regulates the adhesion and signal transduction in T lymphocytes, and the adhesion of thymocytes to thymic stroma. The interaction between 30-H12 and the antibody to the CD3/TCR complex upregulates thymocytes signal transduction and apoptosis and downregulates mature T cell proliferation. The 30-H12 antibody seems to be unable to cross-link with CD90.1.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR