Anti-CD40 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5C3]

Supplier: Biogems
02511-80-100
75842-772EA 637.99 USD
75842-772
Anti-CD40 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5C3]
Antibodies
The 5C3 monoclonal antibody specifically reacts with human CD40, a 48kDA type I glycoprotein. CD40 is part of the TNFR family and expressed on a subset of T cells, B cells, dendritic cells, and macrophages. It has been shown to be involved in the protection of B cells from apoptosis, B cell differentiation, co-stimulation, and isotype class-switching. The 5C3 antibody partially blocks the binding of CD40L (CD154). In the presence of IL-4 or PMA, the 5C3 antibody promotes B cell proliferation.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR