Anti-CD30 Armenian Hamster Monoclonal Antibody [clone: mCD30.1]

Supplier: BIOGEMS INTERNATIONAL INC.
02412-25-500
75842-762EA 703.79 USD
75842-762
Anti-CD30 Armenian Hamster Monoclonal Antibody [clone: mCD30.1]
Antibodies
The mCD301 monoclonal antibody specifically binds to mouse CD30, a member of the Tumor Necrosis Factor Receptor (TNFR) superfamily. It is expressed on activated T, B, NK, and monocyte cells. Its expression on activated T lymphocytes is regulated by cytokines and CD28. The ligand for CD30 is CD153. It has been suggested that the molecule plays a role in the protection against autoimmunity and the regulation of Th1 and Th2 cytokine secretion.

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: mCD30.1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR