Anti-CD30 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Ber-H2]

Supplier: Biogems
02411-60-100
75842-754EA 556.46 USD
75842-754
Anti-CD30 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Ber-H2]
Antibodies
The Ber-H2 monoclonal antibody specifically reacts with human CD30, a 120kDA type I transmembrane glycoprotein of the tumor necrosis factor receptor superfamily (TNFR) also known as the Ki-1 antigen. CD30 can elicit signals leading to either activation or apoptosis through interaction with CD30 ligand (CD30L). It is expressed by a subset of extrafollicular activated B and T cells, lung macrophages, activated NK cells, lymphomas, and endothelial cells. It is highly expressed on Hodgkins and Reed-Sternberg cells.

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: Ber-H2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR