Anti-CD20 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H7]

Supplier: BIOGEMS INTERNATIONAL INC.
02311-60-100
75842-738EA 296.79 USD
75842-738
Anti-CD20 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H7]
Antibodies
The 2H7 monoclonal antibody specifically reacts with human CD20, a 33-37kDA B-lymphocyte surface molecule. CD20 is an unglycosylated four-transmembrane phosphoprotein expressed by B cells in all stage of development, except the final plasma cells. It has been reported that the molecule is involved in B cell activation and has also been observed on a subset of circulating T lymphocytes.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR