Anti-FceR1 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 42795]

Supplier: Biogems
84112-50-100
75842-668EA 187.11 USD
75842-668
Anti-FceR1 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 42795]
The Mar-1 monoclonal antibody binds to the Fc ε Receptor I α subunit (FceR1a), which is a transmembrane glycoprotein from the immunoglobulin superfamily. FceR1a lacks signal-transducing ability and is expressed by mast and basophil cells.The Fc ε Receptor I α subunit is upregulated by IgE and forms a tetramer with a beta subunit and two gamma subunits, which have ITAM (immunoreceptor tyrosine-based activation motifs). The complex formed by the four subunits has very important roles in the IgE-facilitated allergic reactions.

Type: Primary
Antigen: FceR1
Clonality: Monoclonal
Clone: 42795
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR