Anti-Fc epsilon Receptor I alpha (FceR1) (MAR-1) Armenian Hamster Monoclonal Antibody [clone: 42795]

Supplier: Biogems
84112-20-100
75842-662EA 154.85 USD
75842-662
Anti-Fc epsilon Receptor I alpha (FceR1) (MAR-1) Armenian Hamster Monoclonal Antibody [clone: 42795]
Antibodies
The Mar-1 monoclonal antibody binds to the Fc ε Receptor I α subunit (FceR1a), which is a transmembrane glycoprotein from the immunoglobulin superfamily. FceR1a lacks signal-transducing ability and is expressed by mast and basophil cells.The Fc ε Receptor I α subunit is upregulated by IgE and forms a tetramer with a beta subunit and two gamma subunits, which have ITAM (immunoreceptor tyrosine-based activation motifs). The complex formed by the four subunits has very important roles in the IgE-facilitated allergic reactions.

Type: Primary
Antigen: Fc epsilon Receptor I alpha (FceR1) (MAR-1)
Clonality: Monoclonal
Clone: 42795
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR