Anti-Foxp3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MF23]

Supplier: Biogems
83812-60-25
75842-648EA 174.21 USD
75842-648
Anti-Foxp3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MF23]
Antibodies
The MF23 monoclonal antibody specifically reacts with the mouse 50-55 kDa Foxp3 protein (JM2, IPEX), a member of the forkhead family of transcription factors. Foxp3 is expressed by the Treg lymphocytes, whose development and function are influenced by the forkhead protein. Ectopic expression of Foxp3 in T lymphocytes inhibits their activity and cytokine expression. Mutations of Foxp3 result in the “scurfy” mice phenotype. It is reported that the MF23 antibody recognizes an epitope between 1-87 amino acids in the N-terminal domain of mouse Foxp3.

Type: Primary
Antigen: Foxp3
Clonality: Monoclonal
Clone: MF23
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR