NK1.1 (CD161) Monoclonal Antibody 100ug
Supplier: Biogems
NK1.1 (CD161), Monoclonal Antibody, Clone: PK136, Host: Mouse, Species Reactivity: Mouse, Isotype: IgG2a, kappa, Conjugate:PerCP-Cy5.5, Formulation: Phosphate-buffered aqueous solution, pH7.2, Application: Flow Cytometry, Size: 100ug