NK1.1 (CD161) Mab 25ug
Supplier: Biogems
NK1.1 (CD161), Monoclonal Antibody, Clone: PK136, Host: Mouse, Species Reactivity: Mouse, Isotype: IgG2a, kappa, Conjugate:PE, Formulation: Phosphate-buffered aqueous solution,