Anti-Foxp3 (3G3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3G3]

Supplier: BIOGEMS INTERNATIONAL INC.
83412-60-100
75842-602EA 425.85 USD
75842-602
Anti-Foxp3 (3G3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3G3]
Antibodies
The 3G3 monoclonal antibody specifically reacts with the mouse 50-55 kDa Foxp3 protein (JM2, IPEX), a member of the forkhead family of transcription factors. Foxp3 is expressed by the Treg lymphocytes, whose development and function are influenced by the forkhead protein. Ectopic expression of Foxp3 in T lymphocytes inhibits their activity and cytokine expression.Mutations of Foxp3 result in the “scurfy” mice phenotype.

Type: Primary
Antigen: Foxp3 (3G3)
Clonality: Monoclonal
Clone: 3G3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR