Anti-IL-12/IL-23 p40 (C17.8) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: C17.8]

Supplier: Biogems
83312-60-25
75842-596EA 103.23 USD
75842-596
Anti-IL-12/IL-23 p40 (C17.8) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: C17.8]
The C17.8 antibody specifically reacts with the p40 subunit of IL-12, as free monomer, as a homodimer, or as a part of the p70 dimer. C17.8 seems to neutralize the activity of mouse IL-12 and IL-23. IL-12 is the p70 heterodimer of p35 and p40, expressed by the macrophages, dendritic cells, and monocytes. The C17.8 antibody cross-reacts with IL-23, since IL-23 also contains one p40 subunit.

Type: Primary
Antigen: IL-12/IL-23 p40 (C17.8)
Clonality: Monoclonal
Clone: C17.8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR