Ly-6G (Gr-1) Mab 100ug
Supplier: Biogems
Ly-6G (Gr-1), Monoclonal Antibody, Clone: RB6-8C5, Host: Rat, Species Reactivity: Mouse, Isotype: IgG2b, kappa, Conjugate: PE-Cy7, Formulation: Phosphate-buffered aqueous solution,