Anti-Ly-6G Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RB6-8C5]

Supplier: BIOGEMS INTERNATIONAL INC.
83122-60-100
75842-562EA 193.56 USD
75842-562
Anti-Ly-6G Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RB6-8C5]
Antibodies
The RB6-8C5 monoclonal antibody reacts with the mouse Ly-6G (also known as Gr-1). The Ly-6G protein is a myeloid differentiation antigen of 21-25 kDa, expressed in a regulated manner by the myeloid lineage in the bone marrow, where the level of antigen expression is correlated with the granulocyte maturation and differentiation. In the bone marrow, the antigen is not expressed by the erythroid cells. From the peripheric cells, RB6-8C5 binds with monocytes, neutrophils and eosinophils.As a marker for the mouse monocytes, macrophages and granulocytes, the RB6-8C5 antibody is usually combined with M1/70, a macrophage labeling antibody (Anti-CD11b), for phenotypic analysis.

Type: Primary
Antigen: Ly-6G
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR