Anti-OCT3/4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: EM92]

Supplier: Biogems
78011-60-100
75842-544EA 764.34 USD
75842-544
Anti-OCT3/4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: EM92]
Antibodies
The EM92 monoclonal antibody reacts with human and mouse Oct 3/4, a POU family transcription factor. Oct 3/4 is encoded by the Pou5F1 gene and expressed on undifferentiated cells and lost in adult tissue. Fbx15, FGF-4, NANOG, Rex1, and UTF1 expression is regulated with Oct 3/4 and in many cases gene expression is regulated in concert with Sox2.

Type: Primary
Antigen: OCT3/4
Clonality: Monoclonal
Clone: EM92
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR