Anti-GARP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: GARP5]

Supplier: Biogems
77111-80-100
75842-536EA 464.55 USD
75842-536
Anti-GARP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: GARP5]
Antibodies
The GARP5 monoclonal antibody specifically reacts with human Glycoprotein A Repetitions Predominant (GARP, also known as LRRC32 or Garpin) molecule, a 80 kDa transmembrane glycoprotein expressed on the cell surface. GARP is expressed in activated Treg cells, placenta, pancreas, liver, skeletal muscle, heart, lung, and kidney, and it acts as a receptor for latent TGF-β, which down-regulates the Treg cells action. 

Type: Primary
Antigen: GARP
Clonality: Monoclonal
Clone: GARP5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR