GARP Monoclonal Antibody 100tests
Supplier: Biogems
GARP, Monoclonal Antibody, Clone: GARP5, Host: Mouse, Species Reactivity: Human, Isotype: IgG1, Conjugate:PE, Formulation: Phosphate-buffered aqueous solution,