Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LN3]

Supplier: Biogems
74111-60-25
75842-510EA 186.16 USD
75842-510
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LN3]
Antibodies
The LN3 monoclonal antibody specifically reacts with the human leukocyte antigen-DR (HLA-DR), a MHC class II heterodimeric surface glycoprotein. HLA-DR is expressed on the antigen presenting surfaces of monocytes, macrophages, dendritic cells, activated T cells, and B cells. It is integral to peptide presentation to CD4+ T cells.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: LN3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR