HLA-DR Mab 25tests
Supplier: Biogems
HLA-DR, Monoclonal Antibody, Clone: LN3, Host: Mouse, Species Reactivity: Human, Isotype: IgG2b, kappa, Conjugate: FITC, Formulation: Phosphate-buffered aqueous solution,