Anti-IL-17A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17B7]

Supplier: BIOGEMS INTERNATIONAL INC.
73812-60-50
75842-500EA 363.21 USD
75842-500
Anti-IL-17A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17B7]
Antibodies
The 17B7 monoclonal antibody specifically binds to mouse and rat IL-17A, a pro-inflammatory cytokine. It is produced by T helper 17 (Th17) cells, a unique subset of IL-23 dependent CD4+ T cells. Interleukin-17A is highly expressed in transplant rejection, asthma, psoriasis, and multiple sclerosis, and enhances the expression of ICAM-1 in human fibroblasts. The homodimer is expressed by activated peripheral CD4+ T lymphocytes. The Interleukin-17A binds to the IL-17 receptors (IL-17R) expressed by mast cells, monocytes and macrophages, fibroblasts, and endothelial and epithelial cells.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone: 17B7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR