IL-17A Mab 25ug
Supplier: Biogems
IL-17A, Monoclonal Antibody, Clone: 17B7, Host: Rat, Species Reactivity: Mouse, Rat, Isotype: IgG2a, kappa, Conjugate: FITC, Formulation: Phosphate-buffered aqueous solution,