Mouse IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: C1.18.4]

Supplier: Biogems
43912-50-100
75842-488EA 116.13 USD
75842-488
Mouse IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: C1.18.4]
Antibodies
The C1.18.4 monoclonal antibody is used as an Isotype Control for MouseIgG2a antibodies. It has unknown specificity and occurred spontaneously in a C3H mouse.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR