Mouse IgG2a Isotype Control (BG Violet 450) [clone: C1.18.4]

Supplier: BIOGEMS INTERNATIONAL INC.
43912-40-100
75842-486EA 206.45 USD
75842-486
Mouse IgG2a Isotype Control (BG Violet 450) [clone: C1.18.4]
Antibodies
The C1.18.4 monoclonal antibody is used as an Isotype Control for MouseIgG2a antibodies. It has unknown specificity and occurred spontaneously in a C3H mouse. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR