Rat IgG1 Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: HRPN]

Supplier: Biogems
42813-70-100
75842-476EA 219.36 USD
75842-476
Rat IgG1 Isotype Control (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: HRPN]
Antibodies
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR