Rat IgG1 Isotype Control (BG Violet 450) [clone: HRPN]

Supplier: BIOGEMS INTERNATIONAL INC.
42813-40-100
75842-470EA 174.21 USD
75842-470
Rat IgG1 Isotype Control (BG Violet 450) [clone: HRPN]
Antibodies
The HRPN monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG1 antibodies.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: HRPN
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR