Rat IgG2b Isotype Control (APC (Allophycocyanin)) [clone: LTF-2]

Supplier: Biogems
41823-80-100
75842-462EA 200.04 USD
75842-462
Rat IgG2b Isotype Control (APC (Allophycocyanin)) [clone: LTF-2]
Antibodies
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR