Rat IgG2b Isotype Control [clone: LTF-2]

Supplier: BIOGEMS INTERNATIONAL INC.
41823-20-100
75842-444EA 116.13 USD
75842-444
Rat IgG2b Isotype Control [clone: LTF-2]
Antibodies
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR