Rat IgG2a Isotype Control (PE (Phycoerythrin)-Cy5®) [clone: 2A3]

Supplier: BIOGEMS INTERNATIONAL INC.
41813-65-100
75842-434EA 200.04 USD
75842-434
Rat IgG2a Isotype Control (PE (Phycoerythrin)-Cy5®) [clone: 2A3]
Antibodies
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR