Rat IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: 2A3]

Supplier: Biogems
41813-50-100
75842-430EA 129.04 USD
75842-430
Rat IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate)) [clone: 2A3]
Antibodies
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR