Anti-CD278 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ISA-3]

Supplier: Biogems
30811-60-25
75842-392EA 352.01 USD
75842-392
Anti-CD278 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ISA-3]
Antibodies
The ISA-3 monoclonal antibody specifically reacts with human CD278 or Inducible COStimulatory molecule (ICOS), a 47-60 kDA homodimeric glycoprotein also known as H4, AILIM, and CRP-1. It is expressed on activated T cells and a group of thymocytes and is involved in the proliferation, activation, and cytokine production of T cells. When plate-bound, the ISA-3 antibody induces the production of IL-4,5,10 in T cells.

Type: Primary
Antigen: CD278
Clonality: Monoclonal
Clone: ISA-3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR