Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]

Supplier: BIOGEMS INTERNATIONAL INC.
26812-25-500
75842-364EA 283.89 USD
75842-364
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Antibodies
The 10F.9G2 monoclonal antibody specifically reacts with mouse CD274, also known as B7-H1 or PD-L1, a 43 kDa glycoprotein of the B7 family of the immunoglobulin superfamily. CD274 is expressed on the B and T lymphocytes, natural killer cells, and dendritic cells. The receptor for the B7-H1 molecule is PD-1, which contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM), and is expressed on activated B and T cells. The interaction between CD274 and PD-1 seems to downregulate the T and B immune responses.

Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 10F.9G2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR