Anti-CD40LG Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MR1]

Supplier: Biogems
16612-50-100
75842-296EA 290.34 USD
75842-296
Anti-CD40LG Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MR1]
Antibodies
The MR1 monoclonal antibody specifically reacts with mouse CD154, a 39kDA transmembrane glycoprotein also known as CD40 ligand and gp39. CD154 is expressed on activated T cells and plays a role in B-T cell costimulation. The interaction of CD154 with CD40 induces B cell cycle entry and the signaling for T-dependent B cell activation. By interfering with this interaction, the MR1 antibody is reported to inhibit T and B lymphocyte activation.

Type: Primary
Antigen: CD40LG
Clonality: Monoclonal
Clone: MR1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR