Anti-CD144 (VE-Cadherin) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: BV13]

Supplier: Biogems
16512-60-25
75842-288EA 294.1 USD
75842-288
Anti-CD144 (VE-Cadherin) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: BV13]
Antibodies
The BV13 monoclonal antibody specifically reacts with mouse CD144 (VE-Cadherin), a 120 kDA type II Cadherin. CD144 is a hemophilic adhesion molecule of the endothelial cell line essential to maintaining cell layer integrity. It is involved in cell growth inhibition, migration, and survival.

Type: Primary
Antigen: CD144 (VE-Cadherin)
Clonality: Monoclonal
Clone: BV13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR