Anti-CD123 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6H6]

Supplier: BIOGEMS INTERNATIONAL INC.
15311-80-100
75842-272EA 657.91 USD
75842-272
Anti-CD123 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6H6]
Antibodies
The 6H6 monoclonal antibody specifically reacts with human CD123, the alpha chain subunit of the IL-3 receptor. CD123’s affinity to IL-3 is low, but increases when associated with CD131. It can be found on basophils, dendritic cells, macrophages, and mast cells.

Type: Primary
Antigen: CD123
Clonality: Monoclonal
Clone: 6H6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR