Anti-CD103 Armenian Hamster Monoclonal Antibody [clone: 2E7]

Supplier: BIOGEMS INTERNATIONAL INC.
15112-25-100
75842-258EA 448.5 USD
75842-258
Anti-CD103 Armenian Hamster Monoclonal Antibody [clone: 2E7]
Antibodies
The 2E7 monoclonal antibody specifically reacts with mouse CD103, a type I transmembrane glycoprotein in the integrin family that bind to E-cadherin. CD103 is expressed on intraepithelial lymphocyte T cells, peripheral regulatory T cells, lamina propria T cells, and a subset of dendritic cells. It is reported to be involved in T cell homing and immune system regulation.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: 2E7
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR