Anti-CD172a Rat Monoclonal Antibody [clone: P84]

Supplier: Biogems
14812-25-500
75842-256EA 837.92 USD
75842-256
Anti-CD172a Rat Monoclonal Antibody [clone: P84]
Antibodies
The P84 monoclonal antibody specifically reacts with mouse CD172a, a type I transmembrane protein otherwise known as the signal-regulatory protein alpha. It is an adhesion molecules expressed on dendritic cells, monocytes, macrophages, and neuronal tissue. It is reported to be involved in regulating the phagocytic activity of macrophages and mediating neural synaptic activity. The P84 antibody is reported to have neutralizing capability.

Type: Primary
Antigen: CD172a
Clonality: Monoclonal
Clone: P84
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR