CD152 (CTLA-4) Monoclonal Antibody 25ug
Supplier: Biogems
CD152 (CTLA-4), Monoclonal Antibody, Clone: UC10-4F10-11, Host: Armenian Hamster, Species Reactivity: Mouse, Isotype: IgG, Conjugate:APC, Formulation: Phosphate-buffered aqueous solution, pH7.2, Application: Flow Cytometry, Store at 4 deg C, Size: 25ug