Anti-CTLA4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BNI3]

Supplier: Biogems
14611-80-25
75842-236EA 148.4 USD
75842-236
Anti-CTLA4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BNI3]
Antibodies
The BNI3 monoclonal antibody specifically reacts with human CD152, the Cytotoxic T-Lymphocyte Antigen 4 (CTLA-4). CTLA-4 is expressed on activated CD28+ T cells, and binds the B7 family members B7-1 (CD80) and B7-2 (CD86). The structure of CTLA-4 is similar to the structure of CD28, but the two molecules seem to have opposite roles on the T lymphocytes. CTLA-4 inhibits the progression of T cell activation, while CD28 stimulates it. This result explains the stimulating role that the immobilization of BNI3 plays on the T lymphocytes proliferation induced by CD28.

Type: Primary
Antigen: CTLA4
Clonality: Monoclonal
Clone: BNI3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR