Anti-CD69 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: FN50]

Supplier: Biogems
11711-50-100
75842-212EA 403.36 USD
75842-212
Anti-CD69 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: FN50]
Antibodies
The FN50 monoclonal antibody specifically reacts with human CD69, the 27-33 kDA type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR