Anti-CD59 (Protectin) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OV9A2]

Supplier: BIOGEMS INTERNATIONAL INC.
11611-60-25
75842-210EA 217.68 USD
75842-210
Anti-CD59 (Protectin) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OV9A2]
Antibodies
The OV9A2 monoclonal antibody specifically reacts with human CD59 (Protectin), a 19-25 kDA GPI-anchored glycoprotein. CD59 is expressed on hematopoietic and non-hematopoietic cells including lymphocytes, granulocytes, erythrocytes, monocytes and neurons. CD59 can inhibit complement cytolytic activity and the assembly of the membrane attack complex when bound to C8 and C9 complement components. It is involved in T cell activation and modulates T cell adhesion through interaction with CD2.

Type: Primary
Antigen: CD59 (Protectin)
Clonality: Monoclonal
Clone: OV9A2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR